Order Entry
Netherlands
ContactUsLinkComponent
 
 
2-Chloro-4,6-dimethoxy-1,3,5-triazine ≥97.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
 
2-Chloro-4,6-dimethoxy-1,3,5-triazine ≥97.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Argentmetric Titration):
    min. 97.0 %
  • Melting point:
    76.0 to 80.0 deg-C

 

Frequently Bought Together