Order Entry
Netherlands
ContactUsLinkComponent
2,4-Toluenediisocyanate ≥98.0% (by GC)
Toxic
Health
2,4-Toluenediisocyanate ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Toxic
Health
2,4-Toluenediisocyanate ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White or Colorless powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together