Order Entry
Netherlands
ContactUsLinkComponent
 
 
Oxamic acid ≥97.0% (by total nitrogen and titration analysis)
 :  TCI
 
undefined
 
Oxamic acid ≥97.0% (by total nitrogen and titration analysis)
 :  TCI

 

  • Appearance:
    White powder to crystal
  • Purity (Neutralization titration):
    min. 97.0 %
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Solubility in hot Water:
    almost transparency

 

Frequently Bought Together