Order Entry
Netherlands
ContactUsLinkComponent
 
 
2-Hydroxy-5-methylpyridine ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
2-Hydroxy-5-methylpyridine ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    182.0 to 186.0 deg-C

 

Frequently Bought Together