
2-Bromo-1,4-difluorobenzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:399-94-0
:
Formula:C₆H₃BrF₂
MDL Number:MFCD00000345
:192.99 g/mol
Boiling Pt:58°C
Melting Pt:–32 °C
Density:1,7414 g/cm³ (20 °C)
Flash Pt:65°C
EINECS:206-920-0
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together