
3,5-Dibromo-1-fluorobenzene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1435-51-4
:
Formula:C₆H₃Br₂F
MDL Number:MFCD00061119
:253.9 g/mol
Boiling Pt:84°C
Density:2,01 g/cm³ (20 °C)
:GHS07
Specification Test Results
- Appearance:Colorless to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together