
5-Bromo-2-chlorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:21739-92-4
:
Formula:C₇H₄BrClO₂
MDL Number:MFCD00002415
:235.46 g/mol
Boiling Pt:316 °C (1013 hPa)
Melting Pt:157…160 °C
Storage Temperature:Ambient
EINECS:244-558-5
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:157.0 to 160.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together