
3,5-Dibromo-4-hydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0189-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:182.0 to 186.0 deg-C
Specification Test Results
About this item
:
:279.92 g/mol
EINECS:221-017-1
:GHS07
CAS Number:2973-77-5
Formula:C₇H₄Br₂O₂
MDL Number:MFCD00016980