

3,5-Dibromosalicylaldehyde ≥98.0% (by HPLC, titration analysis)
: TCI
:90-59-5
:
:C₇H₄Br₂O₂
:MFCD00003318
:279.92 g/mol
:14,03027
:261 °C (1013 hPa)
:86 °C
:202-003-4
:3077
:9,III
:GHS07
:GHS09
- Appearance:White to Orange to Green powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:81.0 to 85.0 deg-C
Frequently Bought Together