4-Bromo-3,5-dimethoxybenzaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:31558-40-4
:
Formula:C₉H₉BrO₃
MDL Number:MFCD00040799
:245.07 g/mol
EINECS:250-698-8
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:113.0 to 117.0 deg-C
- Acid Value:max. 1.0
Specification Test Results
Frequently Bought Together