
1,3-Dibromobenzene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:108-36-1
:
Formula:C₆H₄Br₂
MDL Number:MFCD00000078
:235.91 g/mol
Boiling Pt:218 °C (1013 hPa)
Melting Pt:–7 °C
Density:1,952 g/cm³ (20 °C)
Flash Pt:94°C
EINECS:203-574-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together