
3-Hydroxy-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:602-00-6
:
Formula:C₇H₅NO₅
MDL Number:MFCD00017003
:183.12 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:180.0 to 184.0 deg-C
Specification Test Results
Frequently Bought Together