Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,4-Bis(trimethylsilyl)-1,3-butadiyne ≥99.0% (by GC)
 :  TCI
 
undefined
 
1,4-Bis(trimethylsilyl)-1,3-butadiyne ≥99.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    110.0 to 113.0 deg-C

 

Frequently Bought Together