Order Entry
Netherlands
ContactUsLinkComponent
2-Hydroxyethyl methacrylate ≥95.0% (by GC) stabilised
Irritant
2-Hydroxyethyl methacrylate ≥95.0% (by GC) stabilised
Supplier:  TCI
Certificates
undefined
Irritant
2-Hydroxyethyl methacrylate ≥95.0% (by GC) stabilised
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

Related Information

Frequently Bought Together