

1-Octyne ≥95.0% (by GC)
: TCI
:629-05-0
:
:C₈H₁₄
:MFCD00009546
:126°C
:–1 °C
:0,7461 g/cm³ (20 °C)
:17°C
:Ambient
:211-069-3
:3,II
:110.2 g/mol
:3295
:GHS02
:GHS08
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together