
1-Dodecyne ≥95.0% (by GC)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:765-03-7
:
Formule:C₁₂H₂₂
MDL-nummer:MFCD00008960
:166.31 g/mol
Kokend Pt:100°C
Smelten Pt:–19 °C
Dichtheid:0,778 g/cm³ (20 °C)
Flits Pt:79°C
EINECS:212-134-9
:GHS05
Specificatie testresultaten
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specificatie testresultaten
Frequently Bought Together