

2,3-Dimethyl-2-butene ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:563-79-1
:
Formula:C₆H₁₂
MDL Number:MFCD00008897
:84.16 g/mol
Boiling Pt:73°C
Melting Pt:–75 °C
Density:0,7188 g/cm³ (20 °C)
Flash Pt:–16°C
Storage Temperature:Refrigerator
EINECS:209-263-8
UN:3295
ADR:3,II
:GHS02
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together