


1-Heptene ≥98.0% (by GC)
: TCI
:592-76-7
:
:C₇H₁₄
:MFCD00009531
:98.19 g/mol
:94°C
:–119 °C
:0,7 g/cm³ (20 °C)
:–4°C
:Ambient
:209-767-8
:2278
:3,II
:GHS02
:GHS07
:GHS08
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together