
1,1-Bis(methylthio)-2-nitroethylene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:13623-94-4
:
Formula:C₄H₇NO₂S₂
MDL Number:MFCD00010443
:165.24 g/mol
Melting Pt:122…125 °C
Storage Temperature:Refrigerator
EINECS:237-108-4
ADR:6.1,II
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:124.0 to 128.0 deg-C
Specification Test Results
Frequently Bought Together