2,4-Dimethylpentane ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:108-08-7
:
Formula:C₇H₁₆
MDL Number:MFCD00008945
Boiling Pt:81
Molecular Weight:100.2 g/mol
Melting Pt:-119.2
Flash Pt:–12°C
EINECS:203-548-0
UN:1206
ADR:3 II
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together