Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
6-Methyl-2,4-heptanedione ≥97.0% (by GC)
  TCIAM0597-5ML
 :  TCI
 :  
undefined
 
 
6-Methyl-2,4-heptanedione ≥97.0% (by GC)
  TCIAM0597-5ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :142.2 g/mol
 :183°C
 :0,92 g/cm³ (20 °C)
 :59°C
 :Refrigerator
 :221-093-6
 :1224
 :3,III
 :GHS02
 :GHS07
 :3002-23-1
 :C₈H₁₄O₂
 :MFCD00008939