Order Entry
Netherlands
ContactUsLinkComponent
 
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol ≥98.0% (by GC)
 :  TCI
 
undefined
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    66.0 to 70.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

Frequently Bought Together