Order Entry
Netherlands
ContactUsLinkComponent
1H,1H-Heptadecafluoro-1-nonanol ≥97.0% (by GC)
1H,1H-Heptadecafluoro-1-nonanol ≥97.0% (by GC)
Catalog # TCIAH1232-5G
Supplier:  TCI
CAS Number:  
undefined
1H,1H-Heptadecafluoro-1-nonanol ≥97.0% (by GC)
Catalog # TCIAH1232-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    67.0 to 71.0 deg-C

Specification Test Results

About this item

 :
Boiling Pt:176°C
 :450.1 g/mol
CAS Number:423-56-3
Formula:C₉H₃F₁₇O
MDL Number:MFCD00153183