Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,4-Dibromooctafluorobutane ≥98.0% (by GC)
  TCIAD3573-5G
 :  TCI
 :  
undefined
 
1,4-Dibromooctafluorobutane ≥98.0% (by GC)
  TCIAD3573-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :104°C
 :359.84 g/mol
 :GHS07
 :335-48-8
 :C₄Br₂F₈
 :MFCD00153112