Order Entry
Netherlands
ContactUsLinkComponent
 
 
trans,trans-1,4-Diphenyl-1,3-butadiene ≥99.0% (by GC)
 :  TCI
 
undefined
 
trans,trans-1,4-Diphenyl-1,3-butadiene ≥99.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Green powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    152.0 to 154.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

 

Frequently Bought Together