Order Entry
Netherlands
ContactUsLinkComponent
1,8-Octanedithiol ≥97.0% (by GC, titration analysis)
Irritant
1,8-Octanedithiol ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
1,8-Octanedithiol ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Iodometric Titration):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together