
1,5-Pentanedithiol ≥95.0% (by GC, titration analysis)
Catalog # TCIAP1518-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
- Purity (Iodometric Titration):min. 95.0 %
Specification Test Results
About this item
:
:136.28 g/mol
Boiling Pt:108°C
Melting Pt:-72°C
Melting Pt:–72°C
Density:1,016 g/cm³ (20 °C)
Flash Pt:95°C
Storage Temperature:Ambient
EINECS:213-194-9
:GHS07
CAS Number:928-98-3
Formula:C₅H₁₂S₂
MDL Number:MFCD00004908