

1,2-Propanedithiol ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:814-67-5
:
Formula:C₃H₈S₂
MDL Number:MFCD00022073
:108.23 g/mol
Flash Pt:35°C
EINECS:212-398-5
UN:1228
ADR:3,III
:GHS02
:GHS06
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
- Specific gravity (20/20):1.0600 to 1.1000
Specification Test Results
Frequently Bought Together