
1-Tetradecanethiol ≥97.0% (by GC)
Catalog # TCIAT0082-25ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
:230.46 g/mol
Boiling Pt:310°C
Melting Pt:7 °C
Density:0,8641 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:218-209-2
ADR:9,
:GHS07
CAS Number:2079-95-0
Formula:C₁₄H₃₀S
MDL Number:MFCD00022099