

1,3-Dichloro-2-propanol ≥98.0% (by GC)
Catalog # TCIAD0402-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:128.99 g/mol
Merck Index:14,03074
Boiling Pt:174°C
Melting Pt:-4°C
Melting Pt:–4°C
Density:1,36 g/cm³ (20 °C)
Flash Pt:74°C
Storage Temperature:Refrigerator
EINECS:202-491-9
UN:2750
ADR:6.1,II
:GHS06
:GHS08
CAS Number:96-23-1
Formula:ClH₂CCHOHCH₂Cl
MDL Number:MFCD00000951