


Dichloromethyl methyl ether ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4885-02-3
:
Formula:C₂H₄Cl₂O
MDL Number:MFCD00000828
:114.96 g/mol
Boiling Pt:85°C
Density:1,29 g/cm³ (20 °C)
Flash Pt:22°C
EINECS:225-498-9
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together