Trichloroacetic acid (TCA) ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Merck Index:14,09627
Boiling Pt:198°C
Melting Pt:58°C
Density:1,63 g/cm³ (20 °C)
Flash Pt: (closed cup)
Storage Temperature:Refrigerator
EINECS:200-927-2
UN:1839
ADR:8,II
:GHS05
:GHS08
:GHS09
Specification Test Results
- Appearance:White powder to crystal
- Purity (Neutralization titration):min. 99.0 % (after drying)
- Solubility in Water:almost transparency
- Drying loss:max. 2.0 %
Specification Test Results
Frequently Bought Together