


Chloroform ≥99.0% (by GC) stabilised for spectrophotometry
Catalog # TCIAC0175-100ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
- Absorbance (250nm):max. 0.500
- Absorbance (260nm):max. 0.150
- Absorbance (270nm):max. 0.050
Specification Test Results
About this item
:
:119.38 g/mol
Merck Index:14,02141
Boiling Pt:60°C
Melting Pt:-64°C
Melting Pt:–64°C
Density:1,4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
:Stabilised with ethanol
REACH:05-2114614906-45
:GHS06
:GHS08
:GHS09
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826