Order Entry
Netherlands
ContactUsLinkComponent
Citric acid ≥98.0% (by titrimetric analysis)
Irritant
Citric acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAC1949-500G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Citric acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAC1949-500G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :192.13 g/mol
Merck Index:14,02326
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149…151 °C
Density:1,665 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
 :GHS07
CAS Number:77-92-9
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669