Potassium propionate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:327-62-8
:
Formula:C₃H₅KO₂
MDL Number:MFCD00045936
:112.17 g/mol
EINECS:206-323-5
Specification Test Results
- Appearance:White powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together