Order Entry
Netherlands
ContactUsLinkComponent
3,4-Hexanedione ≥96.0% (by GC)
Flammable
3,4-Hexanedione ≥96.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
3,4-Hexanedione ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

Frequently Bought Together