

4-Heptanone ≥97.0% (by GC)
: TCI
:123-19-3
:
:C₇H₁₄O
:MFCD00009403
:114.19 g/mol
:14,03345
:143°C
:–34 °C
:0,8116 g/cm³ (20 °C)
:37°C
:Ambient
:204-608-9
:2710
:3,III
:GHS02
:GHS07
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together