
18-Pentatriacontanone ≥95.0% (by GC)
Catalog # TCIAP1290-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:88.0 to 91.0 deg-C
- Solubility in hot Toluene:almost transparency
Specification Test Results
About this item
:
:506.94 g/mol
Boiling Pt:345°C
Melting Pt:89 °C
Density:0,84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-993-1
:GHS07
CAS Number:504-53-0
Formula:C₃₅H₇₀O
MDL Number:MFCD00048491