Order Entry
Netherlands
ContactUsLinkComponent
10-Nonadecanone ≥97.0% (by GC)
10-Nonadecanone ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
10-Nonadecanone ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    58.0 to 62.0 deg-C
  • Solubility in Toluene:
    very faint turbidity

Specification Test Results

Frequently Bought Together