
2-Mercapto-5-methoxybenzimidazole ≥98.0% (by HPLC, titration analysis)
TCIAM1256-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in N,N-DMF:almost transparency
:
:180.23 g/mol
:400 °C (1013 hPa)
:261…263 °C
:253-326-2
:GHS07
:37052-78-1
:C₈H₈N₂OS
:MFCD00134581