3,3'-Diaminobenzidine (DAB) ≥98.0% (by HPLC)
: TCI
: [1,1'-Biphenyl]-3,3',4,4'-tetraamine, DAB, 3,3',4,4'-Biphenyltetramin, Biphenyl-3,3',4,4'-tetrayltetraamine
:91-95-2
:
:C₁₂H₁₄N₄
:MFCD00007725
:214.27 g/mol
:202-110-6
:GHS08
:Refrigerator
- Environmentally Preferable:
- Appearance:White to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:175.0 to 179.0 deg-C
Frequently Bought Together