

3-Amino-9-ethylcarbazool ≥90.0% (by HPLC, titration analysis)
Catalogus # TCIAA1508-5G
Supplier: TCI
CAS Number:
Specifications
- Verpakking:Glass bottle
- VPE:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Brown powder to crystal
- Purity (HPLC):min. 90.0 area%
- Purity (Nonaqueous Titration):min. 90.0 %
Specification Test Results
About this item
:
:210.28 g/mol
Melting Pt:113…114 °C
Storage Temperature:Kamertemperatuur
EINECS:205-057-7
UN:2811
ADR:6.1,III
:GHS06
:GHS08
CAS Number:132-32-1
Formula:C₁₄H₁₄N₂
MDL Number:MFCD00004964