
4,5-Difluoro-o-phenylenediamine ≥97.0% (by GC, titration analysis)
Catalogus # TCIAD3729-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Brown powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:129.0 to 134.0 deg-C
Specification Test Results
About this item
:
:144.12 g/mol
Melting Pt:132°C
:GHS07
CAS Number:76179-40-3
Formula:C₆H₆F₂N₂
MDL Number:MFCD00061131