
Dexamethasone-21-acetate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1177-87-3
:
Formula:C₂₄H₃₁FO₆
MDL Number:MFCD00027407
:434.5 g/mol
Merck Index:14,02943
Melting Pt:240°C
EINECS:214-646-8
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+84.0 to +90.0 deg (C=1, Dioxane)
Specification Test Results
Frequently Bought Together