
3'-Fluoro-4'-methoxyacetophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:455-91-4
:
Formula:C₉H₉FO₂
MDL Number:MFCD00026219
:168.17
Boiling Pt:148°C
Melting Pt:92 °C
EINECS:207-253-8
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:90.0 to 93.0 deg-C
Specification Test Results
Frequently Bought Together