
2,6-Difluoro-4-hydroxybenzonitrile ≥98.0% (by GC, titration analysis)
Catalog # TCIAD3424-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:125.0 to 129.0 deg-C
Specification Test Results
About this item
:
ADR:6.1,III
:155.1 g/mol
UN:3439
:GHS06
CAS Number:123843-57-2
Formula:C₇H₃F₂NO
MDL Number:MFCD03094499