
4-(Trifluoromethoxy)phenylacetonitrile ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:49561-96-8
:
Formula:C₉H₆F₃NO
MDL Number:MFCD00061240
:201.15
Boiling Pt:96°C
Melting Pt:28…30 °C
Density:1,28 g/cm³ (20 °C)
ADR:6.1,III
:GHS07
Specification Test Results
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together