
4-Fluoro-3-methoxybenzonitrile
Supplier: TCI
Certificates
About this item
CAS Number:243128-37-2
:
Formula:C₈H₆FNO
MDL Number:MFCD01569371
:151.14 g/mol
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:108.0 to 111.0 deg-C
Specification Test Results
Frequently Bought Together