
4-(Trifluoromethyl)phenylacetonitrile ≥98.0% (by GC)
Catalog # TCIAT2368-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:45.0 to 49.0 deg-C
Specification Test Results
About this item
:
:185.15 g/mol
Boiling Pt:132°C
Melting Pt:47.85…48.85 °C
Density:1,535 g/cm³ (-150.15 °C)
EINECS:219-052-2
UN:3439
ADR:6.1,III
:GHS06
CAS Number:2338-75-2
Formula:C₉H₆F₃N
MDL Number:MFCD00001921