
3-(Trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:20154-03-4
:
Formula:C₄H₃F₃N₂
MDL Number:MFCD00115018
:136.08 g/mol
Boiling Pt:98°C
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:47.0 to 51.0 deg-C
Specification Test Results
Frequently Bought Together